Key Documents
HAM76008
Hamilton® GC 3-layer septa
diam. × thickness 15.87 mm × 3.81 mm, pack of 12 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
material
laminated silicone rubber
red septa
packaging
pack of 12 ea
parameter
200 °C max. temp.
diam. × thickness
15.87 mm × 3.81 mm
Related Categories
Legal Information
Hamilton is a registered trademark of Hamilton Co.