Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF52905809

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

200mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter

Synonym(s):

NI027150

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 529-058-09

L × wall thickness

200 mm × 0.25 mm

O.D. × I.D.

1.5 mm × 1 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service