Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF30062494

Copper-nickel alloy, Cu70Ni30

200mm tube, 0.4mm outside diameter, 0.15mm wall thickness, 0.1mm inside diameter, hard

Synonym(s):

CU137150

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 300-624-94

L × wall thickness

200 mm × 0.15 mm

O.D. × I.D.

0.4 mm × 0.1 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service