Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D217700

Sigma-Aldrich

1,4-Dithiane

97%

Synonym(s):

(4-Nitrophenyl)acetylene, (4-Nitrophenyl)ethyne, (p-Nitrophenyl)acetylene, 1,4-Dithiacyclohexane, 1-Nitro-4-ethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8S2
CAS Number:
Molecular Weight:
120.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

200 °C (lit.)

mp

107-113 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CSCCS1

InChI

1S/C4H8S2/c1-2-6-4-3-5-1/h1-4H2

InChI key

LOZWAPSEEHRYPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H C Thompson et al.
Journal of chromatography, 467(1), 159-166 (1989-04-21)
1,4-Dithiane is an organo-sulfur chemical associated with production and storage of munitions. Toxicological evaluation of this chemical was scheduled in rats because of the paucity of such information. Prerequisite for the evaluation was the development of gas chromatographic procedures using
H Lee et al.
Mutation research, 321(4), 213-218 (1994-06-01)
1,3-Dithiane and 1,4-dithiane are the sulfur-containing Maillard reaction products (MRPs) which have been found in boiled beef extracts. In this study the genotoxicity of these products was examined using the Salmonella/microsomal test and the CHO/SCE assay. 1,3-Dithiane showed a potent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service