Key Documents
CLS444650
Pyrex® shaker flask with extra-deep baffles
capacity 50 mL
Synonym(s):
Corning® 4446
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
Delong
borosilicate glass
sterility
non-sterile
feature
cap: no
closure type Closure cap
packaging
case of 12
manufacturer/tradename
Corning 4446-50
capacity
50 mL
General description
Delong style neck, with three extra-deep baffles, to enhance gas transfer when used with rotary or reciprocating shakers. Long neck reduces splashing and is designed for two-position polypropylene closures. Closures are sold separately.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.