Skip to Content
MilliporeSigma
All Photos(2)

Documents

C69128

Sigma-Aldrich

3-Chloropropionyl chloride

98%

Synonym(s):

(Chloromethyl)acetyl chloride, 3-Chloropropionic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2COCl
CAS Number:
Molecular Weight:
126.97
Beilstein:
635814
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

143-145 °C (lit.)

density

1.33 g/mL at 25 °C (lit.)

SMILES string

ClCCC(Cl)=O

InChI

1S/C3H4Cl2O/c4-2-1-3(5)6/h1-2H2

InChI key

INUNLMUAPJVRME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Microwave-enhanced liquid-phase synthesis of thiohydantoins and thioxotetrahydropyrimidinones.: This study explores the use of microwave-enhanced synthesis methods for producing thiohydantoins and thioxotetrahydropyrimidinones, with 3-Chloropropionyl chloride as a key reagent. The results indicate that microwave irradiation significantly accelerates the reaction times and enhances the yields, making this a valuable technique for organic synthesis applications (Yeh et al., 2003).
  • Diethylaminopropionamido-hydroxy-anthraquinones as potential anticancer agents: synthesis and characterization.: This paper discusses the synthesis of novel anthraquinone derivatives using 3-Chloropropionyl chloride. These compounds have shown potential as anticancer agents, demonstrating significant biological activity in preliminary tests. The study provides detailed characterization of the synthesized compounds and their potential therapeutic applications (Antonello et al., 1989).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.9 °F - closed cup

Flash Point(C)

70.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service