Key Documents
BR782801
BRAND® insert strips for 24 x 6-well BRANDplates®
Inlet Opening System, PC membrane, size 0.4 μm
Synonym(s):
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
material
PC membrane
description
cellGrade plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
pkg of (individually wrapped)
manufacturer/tradename
BRAND 782801
size
0.4 μm
pore size
0.4 μm
binding type
non-treated surface
Related Categories
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG