Key Documents
BR470816
BRAND® cover glass
L × W 24 mm × 40 mm, rectangular
Synonym(s):
Cover glasses, Microscopy
About This Item
UNSPSC Code:
41122600
NACRES:
NB.16
material
borosilicate glass
rectangular
manufacturer/tradename
BRAND 470816
L × W
24 mm × 40 mm
thickness
0.13-0.17 mm , No. 1
General description
Clear borosilicate glass, hydrolytic class 1, with excellent chemical resistance. Thickness No. 1 (0.13 to 0.17 mm). Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Full automatic production guarantees "ready-for-use" quality - clean, dust and grease free, distortion-free, flatness within ± 3 µm.
Features and Benefits
  • BRAND® borosilicate cover glass offers superior clarity and excellent chemical resistance.
  • Cover glasses for microscope slides are clean, dust/grease free and distortion-free.
  • Full automatic production guarantees "ready-for-use" quality.
  • Flatness within ± 3 μm.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG