Documents
BR152505
BRAND® tubing adapter
high-density polyethylene, for tubing i.d., 8-10 to 11-14 mm
Synonym(s):
Tubing, Tubing adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
high-density polyethylene
feature
for tubing i.d. 8-10 to 11-14 mm
packaging
pack of 10 ea
manufacturer/tradename
BRAND 152505
L
66 mm
General description
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG