Key Documents
BR115015
BRAND® sample vial
for Technicon-Anal., capacity 1.5 mL
Synonym(s):
Sample Cups, Sample vials
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
colorless tube
polystyrene
polystyrene tube
self-standing bottom
sterility
non-sterile
feature
graduations: no
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 115015
capacity
1.5 mL
General description
The BRAND® Sample Vial is designed for secure and efficient storage of samples in laboratory settings. Made from high-quality transparent polystyrene (PS), this vial allows for easy visibility of contents, facilitating quick identification and monitoring.
Features and Benefits
  • BRAND® sample vial is designed for various laboratory applications.
  • Made of transparent polystyrene for accurate and quick sample analysis.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG