Documents
BR114935
BRAND® mini cooler
PC, for 12 tubes
Synonym(s):
Accessories, Microcentrifuge Tubes, Mini cooler, Sample Storage, Tubes
About This Item
UNSPSC Code:
41103025
NACRES:
NB.22
material
PC
description
bench temp.maintained -20°C time 60 min.
feature
for 12 tubes
manufacturer/tradename
BRAND 114935
General description
PC. Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench. Durable polycarbonate filled with non-toxic gel. Mini coolers hold twelve 0.5 ml to 2.0 ml tubes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG