Skip to Content
MilliporeSigma
All Photos(2)

Documents

B74666

Sigma-Aldrich

exo-2-Bromonorbornane

98%

Synonym(s):

exo-2-Bromobicyclo[2.2.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11Br
CAS Number:
Molecular Weight:
175.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5148 (lit.)

bp

82 °C/29 mmHg (lit.)

density

1.363 g/mL at 25 °C (lit.)

SMILES string

Br[C@H]1C[C@@H]2CC[C@H]1C2

InChI

1S/C7H11Br/c8-7-4-5-1-2-6(7)3-5/h5-7H,1-4H2/t5-,6+,7+/m1/s1

InChI key

QXYOAWHKJRWNID-VQVTYTSYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service