Skip to Content
MilliporeSigma
All Photos(2)

Documents

A70909

Sigma-Aldrich

(+)-6-Aminopenicillanic acid

96%

Synonym(s):

(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
Beilstein:
15080
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

powder

optical activity

[α]22/D +276.3°, c = 1.2 in 0.1 M HCl

mp

198-200 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O

InChI

1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1

InChI key

NGHVIOIJCVXTGV-ALEPSDHESA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-6-Aminopenicillanic acid is the core moiety of different penicillins. It can be prepared from the benzylpenicillin silyl ester via cleavage of the amide bond. The coating of (+)-6-aminopenicillanic acid on silver nanoparticles (6APA-AgNPs), imparts them with an ability to detect chromium (IV) in human erythrocytes.
Chemical structure: ß-lactam

Application

(+)-6-Aminopenicillanic acid can be used as a starting material for the synthesis of (6S,7S)-cephalosporins, and organotin polyamine esters by reacting with organotin dihalides via interfacial polycondensation.
It reacts with higher amino acid Schiff bases to form the corresponding β-lactam derivatives.4

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

6-aminopenicillanic acid.
Rolinson GN.
Chemotherapy, 2(1), 52-59 (1961)
Organotin polyamine esters from reaction of 6-aminopenicillanic acid and organotin dihalides
Carraher Jr CE, et al.
J. Polym. Mater., 29(4), 387-387 (2012)
Syntheses of (6S)-cephalosporins from 6-aminopenicillanic acid
Fekner T, et al.
Tetrahedron, 56(33), 6053-6074 (2000)
Anand Pai et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(48), 19810-19815 (2012-11-13)
Bacteria secrete a variety of public good exoproducts into their environment. These exoproducts are typically produced under the control of quorum sensing (QS), a signaling mechanism by which bacteria sense and respond to changes in their density. QS seems to
Zhiqiang Chen et al.
Journal of hazardous materials, 185(2-3), 905-913 (2010-10-26)
A full-scale test was conducted with an up-flow anaerobic sludge blanket (UASB) pre-treating pharmaceutical wastewater containing 6-aminopenicillanic acid (6-APA) and amoxicillin. The aim of the study is to investigate the performance of UASB in the condition of a high chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service