Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96890

Sigma-Aldrich

H-Lys(Z)-OMe hydrochloride

≥97.5% (HPLC)

Synonym(s):

Nε-Z-L-lysine methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OOCNH(CH2)4CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
330.81
Beilstein:
3578476
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.5% (HPLC)

form

powder

optical activity

[α]20/D +14.5±0.5°, c = 2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCc1ccccc1

InChI

1S/C15H22N2O4.ClH/c1-20-14(18)13(16)9-5-6-10-17-15(19)21-11-12-7-3-2-4-8-12;/h2-4,7-8,13H,5-6,9-11,16H2,1H3,(H,17,19);1H/t13-;/m0./s1

InChI key

QPNJISLOYQGQTI-ZOWNYOTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

H-Lys(Z)-OMe hydrochloride also known as Nε-Z-L-lysine methyl ester hydrochloride, is commonly used in solution phase peptide synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service