Skip to Content
MilliporeSigma
All Photos(1)

Documents

95350

Sigma-Aldrich

meso-Tartaric acid monohydrate

≥97%

Synonym(s):

Mesotartaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O6 · H2O
CAS Number:
Molecular Weight:
168.10
Beilstein:
1725146
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021705
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder or crystals

impurities

~10% water

mp

165-166 °C (anhydrous) (lit.)

SMILES string

[H]O[H].O[C@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6.H2O/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);1H2/t1-,2+;

InChI key

UUDLQDCYDSATCH-NUGIMEKKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant invovled in stereoisomeric discrimination in DNA compaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service