Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

932663

Sigma-Aldrich

2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

≥99% (HPLC)

Synonym(s):

Bathocuproine, 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, BCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N2
CAS Number:
Molecular Weight:
360.45
Beilstein:
306714
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.23
Pricing and availability is not currently available.

grade

sublimed grade

Quality Level

description

µe ≈ 5.5 x 10-6 cm2V-1s-1

Assay

≥99% (HPLC)

loss

0.5% TGA, >240°C (weight loss)

mp

279-283 °C (lit.)

solubility

chloroform: soluble
toluene: soluble

λmax

277 nm

fluorescence

λem 386 nm±10 nm in THF

Orbital energy

HOMO 6.4 eV 
LUMO 2.9 eV 

SMILES string

Cc1cc(-c2ccccc2)c3ccc4c(cc(C)nc4c3n1)-c5ccccc5

InChI

1S/C26H20N2/c1-17-15-23(19-9-5-3-6-10-19)21-13-14-22-24(20-11-7-4-8-12-20)16-18(2)28-26(22)25(21)27-17/h3-16H,1-2H3

InChI key

STTGYIUESPWXOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use as Electron Transport / Hole Blocking Layer (ETL / HBL) in OLED and perovskite solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and Stable Perovskite Solar Cells Using Bathocuproine Bilateral-Modified Perovskite Layers
Chen, R. et al.
ACS Applied Materials & Interfaces, 13, 24747-24755 (2021)
Improved color stability in white phosphorescent organic light-emitting diodes using charge confining structure without interlayer
Kim S H, et al.
Applied Physics Letters, 91, 123509-123509 (2007)
Enhanced photovoltage for inverted planar heterojunction perovskite solar cells
Luo, et al.
Science, 360, 1442-1446 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service