MilliporeSigma
930407
OMNI Probe Molybdenum 4-Post Lift-Out Grid
pack of 25 (per vial)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
MA.24
UNSPSC Code:
41111761
Diam.:
3 mm
Packaging:
pack of 25 (per vial)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 25 (per vial)
diam.
3 mm
General description
Custom Mo 4-post lift-out grids for in-situ lift-out. Include multiple indexed mounting locations, two with vertical bars attachment surfaces and two with “V” shaped alignment surfaces. Sides have lower profile for easier access to outermost posts.
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)