Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

923079

Sigma-Aldrich

Copper(II) nitrate hydrate

≥99.99% trace metals basis

Synonym(s):

Cupric nitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · xH2O
CAS Number:
Molecular Weight:
187.56 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

crystals and lumps

mp

114.5 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

VMKYLARTXWTBPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Copper Nitrate hydrate can be used to synthesize novel Cu-BTC composite materials with enhanced water stability and are resitant to change in pH. It is also used as a precursor to prepare Cu2O spherical nanoparticles which can facilitate ligandless synthesis of polyphenylenediethynylenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Composite Materials Resistant to a Change in Medium Acidity from Cu-BTC (MOF-199) and Activated Carbons.
Markova E B, et al.
Petroleum Chemistry, 60(10), 1184-1190 (2020)
Preparation of a composite material AC/Cu-BTC with improved water stability and n-hexane vapor adsorption.
Huang W, et al.
Journal of Nanomaterials (2019)
Copper(i) oxide nanoparticle-mediated C?C couplings for synthesis of polyphenylenediethynylenes: evidence for a homogeneous catalytic pathway.
Pary F, et al.
Catalysis Science & Technology, 11, 2414-2421 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service