919128
Hydroxyapatite/Beta-TCP
composite porous particles (40:60%), 5±1 mm diameter, pore size 400-500 μm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(Ca10(PO4)6(OH)2)4 : (Ca3(PO4)2)6
UNSPSC Code:
12352305
NACRES:
NA.23
Recommended Products
product name
Hydroxyapatite/Beta-TCP composite porous particles (40:60%), 5±1 mm diameter, pore size 400-500 μm
form
solid
Quality Level
H
5 ± 1 mm , cylinder
diameter
5±1 mm
pore size
400-500 μm pore size
Application
Calcium phosphate materials such as hydroxyapatite (HA), α-tricalcium phosphate (α-TCP), and β-tricalcium phosphate (β-TCP) have been widely explored for use in bone regeneration. Composite materials comprising HA and β-TCP incorporate the advantages of both these materials,for enhanced mechanical properties.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hae Lin Jang et al.
Advanced healthcare materials, 5(1), 128-136 (2015-05-13)
Biomimicking ceramics have been developed to induce efficient recovery of damaged hard tissues. Among them, calcium phosphate-based bioceramics have been the most widely used because of their similar composition with human hard tissue and excellent biocompatibilities. However, the incomplete understanding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service