Skip to Content
MilliporeSigma
All Photos(1)

Documents

914835

Sigma-Aldrich

Titania nanoparticles

dispersion, 5 wt. % in ethanol, 150 nm particle size

Synonym(s):

TiO2 nanoparticles, TiOxide, Titanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

5 wt. % in ethanol

particle size

150 nm

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titania nanoparticles are useful for a wide variety of applications, due to their high refractive index and large bandgap, providing optical properties in the visible spectrum. Applications of titania nanoparticles range from energy to medical applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoparticles of Titanium and Zinc Oxides as Novel Agents in Tumor Treatment: a Review.
Bogdan J, et al.
Nanoscale Research Letters, 12(225) (2017)
Modified titanium oxide (TiO2) nanocomposites and its array of applications: a review.
Ghosh S, et al.
Toxicological & Environmental Chemistry, 97:5, 491-514 (2015)
Mini review on photocatalysis of titanium dioxide nanoparticles and their solar applications.
Lan Y, et al.
Nano Energy, 2(5), 1031-1045 (2013)
Introduction: titanium dioxide (TiO2) nanomaterials
Chen X and Selloni A
Chemical Reviews, 114(19), 9281-9282 (2014)

Articles

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service