Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909688

Sigma-Aldrich

Alkyne-A-DSBSO crosslinker

≥95%

Synonym(s):

Alkyne-tagged acid-cleavable disuccinimidyl bissulfoxide, Bis(2,5-dioxopyrrolidin-1-yl) 3,3′-((2-(but-3-yn-1-yl)-2-methyl-1,3-dioxane-5,5-diyl)bis(methylenesulfinyl))dipropionate, Mass spectrometry-cleavable crosslinker for studying protein-protein interactions

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32N2O12S2
CAS Number:
Molecular Weight:
616.66
MDL number:
UNSPSC Code:
12161502

Assay

≥95%

form

powder

availability

available only in USA

storage temp.

2-8°C

SMILES string

[S](=O)(CC3(COC(OC3)(CCC#C)C)C[S](=O)CCC(=O)ON2C(=O)CCC2=O)CCC(=O)ON1C(=O)CCC1=O

InChI key

CWQIIUMERHZYNI-UHFFFAOYSA-N

Application

Alkyne-A-DSBSO (alkyne-tagged, acid-cleavable disuccinimidyl bissulfoxide) Crosslinker is a homobifunctional, membrane-permeable, enrichable disulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS).
Alkyne-A-DSBSO possesses two N-hydroxysuccinimide (NHS) ester groups for targeting amines, a ∼14 Å spacer length, two symmetrical acid-cleavable C-S bonds, and a central bioorthogonal azide tag. After reacting with lysine (Lys) residues, the alkyne tag permits selective enrichment of crosslinked proteins or peptides with a biotin-conjugated azide probe to enhance their detection. Additionally, the post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS.
Alkyne-A-DSBSO Crosslinker has mapped in vivo PPIs for target protein complexes and whole proteomes from living cells. It will also be a useful proteomics tool in the quest for targeting ″undruggable″ protein targets.

Legal Information

Subject to US Patent Application #15/275,001 of the Regents of the University of California

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony M Burke et al.
Organic & biomolecular chemistry, 13(17), 5030-5037 (2015-04-01)
The cross-linking Mass Spectrometry (XL-MS) technique extracts structural information from protein complexes without requiring highly purified samples, crystallinity, or large amounts of material. However, there are challenges to applying the technique to protein complexes in vitro, and those challenges become

Articles

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service