Skip to Content
MilliporeSigma
All Photos(1)

Documents

901595

Sigma-Aldrich

Vinylene carbonate solution

1.0 M in ethyl acetate

Synonym(s):

1,3-Dioxol-2-one, VC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H2O3
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.22

form

liquid

concentration

1.0 M in ethyl acetate

refractive index

n/D 1.377

density

0.915 g/mL

InChI

1S/C3H2O3/c4-3-5-1-2-6-3/h1-2H

InChI key

VAYTZRYEBVHVLE-UHFFFAOYSA-N

Application

Carbonate building block offered as a solution in ethyl acetate for more convenient handling. Vinylene carbonate may also be used as a dienophile in Diels-Alder reactions.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Stomach

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

29.3 °F

Flash Point(C)

-1.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service