Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

852597

Sigma-Aldrich

5-Iodouridine

95%

Synonym(s):

2,4-Dihydroxy-5-iodo-1-β-D-ribofuranosylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O6
CAS Number:
Molecular Weight:
370.10
Beilstein:
33665
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

mp

205-207 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(I)C(=O)NC2=O

InChI

1S/C9H11IN2O6/c10-3-1-12(9(17)11-7(3)16)8-6(15)5(14)4(2-13)18-8/h1,4-6,8,13-15H,2H2,(H,11,16,17)/t4-,5-,6-,8-/m1/s1

InChI key

RKSLVDIXBGWPIS-UAKXSSHOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B L Golden et al.
RNA (New York, N.Y.), 2(12), 1295-1305 (1996-12-01)
For small RNAs, isomorphous heavy-atom derivatives can be obtained by crystallizing synthetic versions that incorporate modified nucleotides such as iodo- or bromouridine. However, such a synthetic approach is not yet feasible for RNAs greater than approximately 40 nt. We have
W T Stump et al.
RNA (New York, N.Y.), 1(1), 55-63 (1995-03-01)
The N-terminal RNA binding domain (RBD) of the human U1A snRNP protein binds tightly and specifically to an RNA hairpin that contains a 10-nucleotide loop. The protein is one of a class of RNA binding proteins that adopts a beta
Dehalogenation reactions in fast atom bombardment mass spectrometry.
S K Sethi et al.
Analytical chemistry, 56(11), 1975-1977 (1984-09-01)
Daniel E Ryan et al.
RNA (New York, N.Y.), 10(8), 1251-1265 (2004-07-24)
Elucidation of the three-dimensional (3D) structures of the two sequential active sites in spliceosomes is essential for understanding the mechanism of premessenger RNA splicing. The mechanism is predicted to be catalyzed by the small nuclear RNA (snRNA) components of spliceosomes.
Michael J Calderon et al.
PloS one, 10(6), e0128942-e0128942 (2015-06-16)
FOXM1, a member of the forkhead box transcription factor family, plays a key role in cell cycling progression by regulating the expression of critical G1/S and G2/M phase transition genes. In vivo studies reveal that Foxm1 null mice have a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service