Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809721

Sigma-Aldrich

Sodium tauroursodeoxycholate-2,2,4,4-d4

≥98 atom % D, ≥98% (CP)

Synonym(s):

Tauroursodeoxycholic-2,2,4,4-d4 acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26D4H40NaNO6S
Molecular Weight:
525.71
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

C[C@H](CCC(NCCS([O-])(=O)=O)=O)[C@H]1CCC2[C@]3([H])[C@@H](O)C[C@]4([H])C([2H])([2H])[C@H](O)C([2H])([2H])C[C@]4(C)[C@@]3([H])CC[C@@]21C.[Na+]

InChI

1S/C26H45NO6S.Na/c1-15(4-7-23(30)27-10-11-34(31,32)33)19-5-6-20-24(19)16(2)12-21-25(20)22(29)14-17-13-18(28)8-9-26(17,21)3;/h15-22,24-25,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t15-,16-,17?,18-,19?,20?,21?,22+,24?,25?,26+;/m1./s1/i8D2,13D2;

InChI key

WRYMNPNETXBBEN-GJDYHZDRSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service