Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803499

Sigma-Aldrich

PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)

Synonym(s):

2,5-Dioxo-1-pyrrolidinyl 17-oxo-19-(2-pyridinyldithio)-4,7,10,13-tetraoxa-16-azanonadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23 H33 N3 O9 S2
CAS Number:
Molecular Weight:
559.65
UNSPSC Code:
12161502
NACRES:
NA.22

form

oil

Quality Level

mol wt

559.65

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

water: soluble

functional group

NHS ester

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCSSC1=CC=CC=N1)NCCOCCOCCOCCOCCC(ON2C(CCC2=O)=O)=O

General description

SPDP-type reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithiol group at the opposite end. PEG4-SPDP has a 4-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.

Features and Benefits

  • Reactive groups: NHS ester and pyrirdyldithiol
  • Efficient amine conjugation—NHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines
  • Reversible sulfhydryl conjugation—pyridyldithiol group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond
  • Polyethylene glycol—PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules
  • Spacer arm—connects primary amines and sulfhydryl targets at distances up to 25.7 angstroms; compare to PEG12-SPDP (54.1 angstroms)

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurath, A. R. and Strick, N.
J. Virol. Methods, 3, 155-165 (1981)
Preparation of antibody-toxin conjugates.
A J Cumber et al.
Methods in enzymology, 112, 207-225 (1985-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service