Skip to Content
MilliporeSigma
All Photos(1)

Documents

77861

Sigma-Aldrich

2-Phenylethanol

≥99.0% (GC)

Synonym(s):

β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1905732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.21 (vs air)

vapor pressure

1 mmHg ( 58 °C)

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.5317 (lit.)

bp

219-221 °C/750 mmHg (lit.)

mp

−27 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenylethanol is an aromatic alcohol used as a flavoring agent in the cosmetic, perfume, and food industries.

Application

  • The bacteriostatic activity of 2-phenylethanol derivatives correlates with membrane binding affinity: This study examines how the bacteriostatic properties of 2-phenylethanol derivatives are influenced by their affinity for binding to membranes, providing insights into their antimicrobial effectiveness (IS Kleinwächter et al., 2021).
  • Anti-depressive-like effect of 2-phenylethanol inhalation in mice: This study explores the neuropsychological effects of inhaling 2-phenylethanol, suggesting potential therapeutic applications for depression (H Ueno et al., 2019).

recommended

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in biotechnological production of 2-phenylethanol
Hua, et al.
Biotechnology Advances, 29, 654-660 (2011)
Advances in 2-phenylethanol production from engineered microorganisms
Wang, et al.
Biotechnology Advances, 37, 403-409 (2019)
Mengwan Li et al.
Biotechnology for biofuels, 14(1), 3-3 (2021-01-08)
2-phenylethanol (2-PE) is a rose-scented flavor and fragrance compound that is used in food, beverages, and personal care products. Compatibility with gasoline also makes it a potential biofuel or fuel additive. A biochemical process converting glucose or other fermentable sugars
Dongliang Hua et al.
Biotechnology advances, 29(6), 654-660 (2011-05-24)
2-Phenylethanol (2-PE) is an important aromatic alcohol with a rose-like fragrance. It has been widely applied in the cosmetic, perfume, and food industries and is mainly produced by chemical synthesis. An alternative method for the production of natural flavors and
Jaspreet Singh et al.
Biochimica et biophysica acta, 1831(4), 747-758 (2013-01-16)
X-linked adrenoleukodystrophy (X-ALD) is a peroxisomal disorder caused by mutations in the ABCD1 gene. Accumulation of very long chain fatty acids (VLCFA) that have been attributed to reduced peroxisomal VLCFA β-oxidation activity are the hallmark of the disease. Overexpression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service