765406
Gold nanoparticles
50 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O
Synonym(s):
Gold nanoparticles NH2 functionalized, Au NP NH2, Gold Colloid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
material
PEG 3000
Quality Level
form
dispersion in H2O
nanoparticles
packaging
glass bottle of 1 mL
OD
50
diameter
50 nm
pH
6.0-8.0 (25 °C)
solubility
water: miscible
density
1.00 g/cm3
λmax
535 nm
functional group
amine
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Amine functionalized, PEG 3000 coated, OD 50, H2O dispersed gold nanoparticles (50 nm) can be used in the activation of silica nanosphere for the decomposition of formic acid. They may be used in the radiosensitization and imaging of cancer cells.
AuNPs can be potentially immobilized on antibodies to improve the sensitivity of optical fiber sensors for the screening of COVID-19. They can also be deposited on the titanium electrode for the formation of a miniaturized device for the electrochemical detection of DNA hybridization, which can be used in the diagnosis of COVID-19.
AuNPs can be potentially immobilized on antibodies to improve the sensitivity of optical fiber sensors for the screening of COVID-19. They can also be deposited on the titanium electrode for the formation of a miniaturized device for the electrochemical detection of DNA hybridization, which can be used in the diagnosis of COVID-19.
Functionalized particles have been extensively used to study cellular uptake of nanoparticles and targeted drug delivery.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gold nanoparticles for radiosensitizing and imaging of cancer cells
Borran AA, et al.
Radiation Physics and Chemistry, 152(25), 137-144 (2018)
Optical Fiber Sensors for Rapid Screening of COVID-19
Nag P, et al.
Transactions of the Indian National Academy of Engineering, 152(25), 1-4 (2020)
Strong metal-molecular support interaction (SMMSI): amine-functionalized gold nanoparticles encapsulated in silica nanospheres highly active for catalytic decomposition of formic acid
Yadav M, et al.
Journal of Materials Chemistry, 22(25), 12582-12586 (2012)
Scott H Brewer et al.
Langmuir : the ACS journal of surfaces and colloids, 21(20), 9303-9307 (2005-09-21)
The interaction of bovine serum albumin (BSA) with gold colloids and surfaces was studied using zeta-potential and quartz crystal microbalance (QCM) measurements, respectively, to determine the surface charge and coverage. The combination of these two measurements suggests that BSA binding
Functionalized gold nanoparticles and their biomedical applications
Tiwari, P. M., Vig, K., Dennis, V. A., & Singh, S. R.
Nanomaterials, 1(1), 31-63 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service