Skip to Content
MilliporeSigma
All Photos(1)

Documents

762326

Sigma-Aldrich

Di-tert-butyl phosphite

95%

Synonym(s):

Di-tert-butyl phosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19O3P
CAS Number:
Molecular Weight:
194.21
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.419

density

0.960 g/L at 25 °C

storage temp.

2-8°C

SMILES string

O=P(OC(C)(C)C)OC(C)(C)C

InChI

1S/C8H19O3P/c1-7(2,3)10-12(9)11-8(4,5)6/h12H,1-6H3

InChI key

HLMXUWAUCZMWOG-UHFFFAOYSA-N

General description

Di-tert-butyl phosphite can be used as a reactant in the synthesis of di-tert-butyl potassium phosphate and di-tert-butyl (chloromethyl) phosphate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

325.4 °F

Flash Point(C)

163 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of efficient processes for the preparation of di-tert-butyl potassium phosphate and di-tert-butyl (chloromethyl) phosphate
Zheng B, et al.
Organic Process Research & Development, 18(5), 636-642 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service