Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760390

Sigma-Aldrich

3-Bromo-1-methyl-1H-pyrazole

96%

Synonym(s):

1-Methyl-3-bromopyrazole, 3-Bromo-1-methylpyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5BrN2
CAS Number:
Molecular Weight:
161.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.528

bp

204-210 °C/760 mmHg

density

1.585 g/mL at 25 °C

SMILES string

CN1C=CC(Br)=N1

InChI

1S/C4H5BrN2/c1-7-3-2-4(5)6-7/h2-3H,1H3

InChI key

ZGEVJEQMVRIEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.0 °F

Flash Point(C)

106.66 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service