Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749648

Sigma-Aldrich

2-[2-(Boc-amino)ethoxy]ethanol

97%

Synonym(s):

2-(2-tert-Butoxycarbonylaminoethoxy)ethanol, N-[2-(2-Hydroxyethoxy)ethyl]carbamic acid tert-butyl ester, tert-Butyl[2-(2-hydroxyethoxy)ethyl]carbamate, [2-(2-Hydroxyethoxy)ethyl]carbamic acid 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.454

density

1.061 g/mL at 25 °C

functional group

amine
ether
hydroxyl

SMILES string

CC(C)(C)OC(=O)NCCOCCO

InChI

1S/C9H19NO4/c1-9(2,3)14-8(12)10-4-6-13-7-5-11/h11H,4-7H2,1-3H3,(H,10,12)

InChI key

KSFVNEXYCULLEJ-UHFFFAOYSA-N

Related Categories

Application

2-[2-(Boc-amino)ethoxy]ethanol is N-terminal protected 2-(2-aminoethoxy)ethanol. It is generally used as a molecular spacer/linker. Some of such applications are:
  • Synthesis of biotinylated HaloLigands to prepare quantum dot adducts for HaloTag protein-mediated specific labeling of live cells.
  • It is one of the key precursors for the total synthesis of tyroscherin and photo-activatable tyroscherin-based affinity reagent used for identification of tyroscherin binding proteins.
  • It can be used to link HaloTag enzyme with fluorescent zinc indicator, ZIMIR to monitor the dynamics of regulated secretion of zinc in real time.
  • It can also be used to synthesize amphiphiles for DNA and siRNA delivery.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis and biological evaluation of tyroscherin.
Tae H S, et al.
Organic Letters, 12(19), 4308-4311 (2010)
Genetic targeting of a small fluorescent zinc indicator to cell surface for monitoring zinc secretion.
Li D, et al.
ACS Chemical Biology, 10(4), 1054-1063 (2015)
HaloTag protein-mediated specific labeling of living cells with quantum dots.
So M K, et al.
Biochemical and Biophysical Research Communications, 374(3), 419-423 (2008)
The effect of charge-reversal amphiphile spacer composition on DNA and siRNA delivery.
Zhang X X, et al.
Bioconjugate Chemistry, 21(5), 988-993 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service