Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741493

Sigma-Aldrich

1,2-Diamino-3,5-dimethylbenzene

96%

Synonym(s):

3,5-Dimethyl-1,2-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

73-78 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(N)c(N)c1

InChI

1S/C8H12N2/c1-5-3-6(2)8(10)7(9)4-5/h3-4H,9-10H2,1-2H3

InChI key

DMEPVFSJYHJGCD-UHFFFAOYSA-N

Application

1,2-Diamino-3,5-dimethylbenzene can be used as a starting material to synthesize:
  • Tetramethyl quinoxaline derivatives by reacting with 1,2-dicarbonyl compounds.
  • 5,7-Dimethyl-1H-benzotriazole by reacting with tert-butyl nitrite.
  • Dimethyl-1H-benzimidazole by reacting with dimethylformamide (DMF).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Expedient synthesis of benzimidazoles using amides
Kattimani PP, et al.
Royal Society of Chemistry Advances, 5(37), 29447-29455 (2015)
tert-Butyl nitrite mediated nitrogen transfer reactions: synthesis of benzotriazoles and azides at room temperature
Azeez S, et al.
Organic & Biomolecular Chemistry, 16(37), 8280-8285 (2018)
A Facile, Catalyst-Free Mechano-Synthesis of Quinoxalines and their In-Vitro Antibacterial Activity Study
Bhutia ZT, et al.
ChemistrySelect, 2(3), 1183-1187 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service