Skip to Content
MilliporeSigma
All Photos(3)

Documents

739103

Sigma-Aldrich

Ru-MACHO®

Synonym(s):

Carbonylchlorohydrido{bis[2-(diphenylphosphinomethyl)ethyl]amino}ethyl]amino}ruthenium(II), {Bis[2-(diphenylphosphino)ethyl]amine}carboynlchlorohydridoruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H30ClNOP2Ru
CAS Number:
Molecular Weight:
607.03
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

260-290 °C

SMILES string

Cl[Ru].[C-]#[O+].C(CP(c1ccccc1)c2ccccc2)NCCP(c3ccccc3)c4ccccc4

InChI

1S/C28H29NP2.CO.ClH.Ru/c1-5-13-25(14-6-1)30(26-15-7-2-8-16-26)23-21-29-22-24-31(27-17-9-3-10-18-27)28-19-11-4-12-20-28;1-2;;/h1-20,29H,21-24H2;;1H;/q;;;+1/p-1

InChI key

GQFWSAKCCNATEV-UHFFFAOYSA-M

Application

<ul>
<li><strong>Dehydrogenative Intramolecular Macrolactonization:</strong> Ru-MACHO catalyst is applied in dehydrogenative intramolecular macrolactonization of dihydroxy compounds, demonstrating its efficiency in organic synthesis with potential implications for pharmaceutical synthesis (Jamdade et al., 2022).</li>
<li><strong>Acceptorless Dehydrogenation of Methanol:</strong> Employing Ru-MACHO catalyst in the acceptorless dehydrogenation of methanol to produce carbon monoxide and hydrogen, this process highlights Ru-MACHO′s capability in catalytic transformations and green chemistry applications (Kaithal et al., 2021).</li>
<li><strong>Precapture and Hydrogenation of CO2:</strong> A study on the heterogenized Ru-MACHO catalytic systems for pre-capture and hydrogenation of CO2 into methanol, underscoring the catalyst′s potential in sustainable chemical processes (Szego et al., 2023).</li>
<li><strong>Pressure-dependent Hydrogenative Depolymerization:</strong> The Ru-MACHO catalyst is used for highly selective pressure-dependent hydrogenative depolymerization of polybutylene succinate, demonstrating the catalyst′s versatility in polymer processing and chemical recycling (Johnson et al., 2024).</li>
</ul>

Legal Information

Sold in collaboration with Takasago for research purposes only. Ru-MACHO is a registered trademark of Takasago Intl. Corp. PCT/JP2010-004301.
Ru-MACHO is a registered trademark of Takasago Intl. Corp.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service