Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

736899

Sigma-Aldrich

Ethyl 2-methyl-2,3-butadienoate

Synonym(s):

2-Methyl-2,3-butadienoic acid ethyl ester, Ethyl 1-methylallene-1-carboxylate, Ethyl 2,3-butadiene-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.459

density

0.954 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)=C=C

InChI

1S/C7H10O2/c1-4-6(3)7(8)9-5-2/h1,5H2,2-3H3

InChI key

QFTBEYHBIFWGHJ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service