Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734403

Sigma-Aldrich

D-Glucose-6-13C,6,6-d2

98 atom % D, 99 atom % 13C, 98% (CP)

Synonym(s):

Deuterated Glucose, Labeled Glucose, Dextrose-6-13C,6,6-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5D2H10O6
CAS Number:
Molecular Weight:
183.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

98% (CP)

form

powder

mp

150-152 °C (lit.)

mass shift

M+3

SMILES string

[2H][13C]([2H])(O)[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1+1D2

InChI key

WQZGKKKJIJFFOK-AUJTXTGDSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Chen et al.
The Journal of clinical endocrinology and metabolism, 100(11), 4082-4091 (2015-09-18)
Whereas insulin resistance and obesity coexist, some obese individuals remain insulin sensitive. We examined phenotypic and metabolic factors associated with insulin sensitivity in both muscle and liver in obese individuals. Sixty-four nondiabetic obese adults (29 males) underwent hyperinsulinemic (15 and
Agnieszka Sitarska et al.
Journal of biomolecular NMR, 62(2), 191-197 (2015-05-02)
For a wide range of proteins of high interest, the major obstacle for NMR studies is the lack of an affordable eukaryotic expression system for isotope labeling. Here, a simple and affordable protocol is presented to produce uniform labeled proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service