Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727520

Sigma-Aldrich

2-(Bromomethyl)quinoline

≥96.5% (GC)

Synonym(s):

α-Bromoquinaldine, 2-Quinolinylmethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8BrN
CAS Number:
Molecular Weight:
222.08
Beilstein:
115410
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (GC)

form

powder

SMILES string

BrCc1ccc2ccccc2n1

InChI

1S/C10H8BrN/c11-7-9-6-5-8-3-1-2-4-10(8)12-9/h1-6H,7H2

InChI key

NNAYPIDFVQLEDK-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service