Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

715794

Sigma-Aldrich

1-(3-Aminopropyl)-2-methyl-1H-imidazole

96%

Synonym(s):

3-(2-Methyl-1H-imidazol-1-yl)propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13N3
CAS Number:
Molecular Weight:
139.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.517

density

1.028 g/mL at 25 °C

SMILES string

Cc1nccn1CCCN

InChI

1S/C7H13N3/c1-7-9-4-6-10(7)5-2-3-8/h4,6H,2-3,5,8H2,1H3

InChI key

NDUDHWBKFFJGMA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service