Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

707511

Sigma-Aldrich

(1R,2R)-(+)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethanediamine

97%

Synonym(s):

(1R,2R)-(+)-N,N′-Dimethyl-1,2-bis[(3-trifluoromethyl)phenyl]-1,2-ethanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18F6N2
CAS Number:
Molecular Weight:
376.34
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]22/D +18.0°, c = 1 in chloroform

mp

112-117 °C

SMILES string

CN[C@@H]([C@H](NC)c1cccc(c1)C(F)(F)F)c2cccc(c2)C(F)(F)F

InChI

1S/C18H18F6N2/c1-25-15(11-5-3-7-13(9-11)17(19,20)21)16(26-2)12-6-4-8-14(10-12)18(22,23)24/h3-10,15-16,25-26H,1-2H3/t15-,16-/m1/s1

InChI key

SBGOGHODWUZHIY-HZPDHXFCSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service