Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

692808

Sigma-Aldrich

(R,R)-DACH-phenyl Trost ligand

95%

Synonym(s):

(1R,2R)-(+)-1,2-Diaminocyclohexane-N,N′-bis(2-diphenylphosphinobenzoyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H40N2O2P2
CAS Number:
Molecular Weight:
690.75
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]20/D +131°, c = 1 in methanol

mp

136-142 °C

SMILES string

O=C(N[C@@H]1CCCC[C@H]1NC(=O)c2ccccc2P(c3ccccc3)c4ccccc4)c5ccccc5P(c6ccccc6)c7ccccc7

InChI

1S/C44H40N2O2P2/c47-43(37-27-13-17-31-41(37)49(33-19-5-1-6-20-33)34-21-7-2-8-22-34)45-39-29-15-16-30-40(39)46-44(48)38-28-14-18-32-42(38)50(35-23-9-3-10-24-35)36-25-11-4-12-26-36/h1-14,17-28,31-32,39-40H,15-16,29-30H2,(H,45,47)(H,46,48)/t39-,40-/m1/s1

InChI key

AXMSEDAJMGFTLR-XRSDMRJBSA-N

Application

Asymmetirc allylic alkylation ligand.

Legal Information

Sold in collaboration with Dr. Reddy′s Laboratories for research purposes only. US Patent 5739396 applies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trost, B.M. et al.
Journal of the American Chemical Society, 124, 11616-11616 (2004)
Asymmetric Transition Metal-Catalyzed Allylic Alkylations.
Barry M. Trost et al.
Chemical reviews, 96(1), 395-422 (1996-02-01)
Trost, B.M. et al.
Aldrichimica Acta, 40, 59-59 (2007)
Asymmetric transition-metal-catalyzed allylic alkylations: applications in total synthesis.
Barry M Trost et al.
Chemical reviews, 103(8), 2921-2944 (2003-08-14)

Articles

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service