Skip to Content
MilliporeSigma
All Photos(3)

Documents

689858

Sigma-Aldrich

2-Bromo-4-nitrophenol

≥98.0%

Synonym(s):

4-Hydroxy-3-bromonitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrNO3
CAS Number:
Molecular Weight:
218.00
Beilstein:
2441518
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)
≥98.0%

form

powder

mp

111-115 °C

SMILES string

Oc1ccc(cc1Br)[N+]([O-])=O

InChI

1S/C6H4BrNO3/c7-5-3-4(8(10)11)1-2-6(5)9/h1-3,9H

InChI key

DCIPFSYBGTWYCR-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service