Skip to Content
MilliporeSigma
All Photos(1)

Documents

687243

Sigma-Aldrich

Buchwald Ligands Kit II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12350000
NACRES:
NA.22

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

functional group

phosphine

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 638439JohnPhos, 97% 1 gSDS

  • 638099CyJohnPhos, 97% 1 gSDS

  • 675938Me4tButylXphos, 96% 250 mgSDS

  • 638080tBuXPhos, 98% 1 gSDS

  • 663131RuPhos, 98% 1 gSDS

  • 638072SPhos, 98% 1 gSDS

  • 638021DavePhos, 97% 1 gSDS

  • 638064XPhos, 98% 1 gSDS

  • 677280sSPhos 500 mgSDS

See All (9)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Buchwald phosphine ligands for C-C, C-N, and C-O bond formation.

Related Content

The Buchwald group has developed a series of highly active and versatile palladium precatalysts and biarylphosphine ligands used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. The ligands are electron-rich, and highly tunable to provide catalyst systems with a diverse scope, high stability and reactivity. Furthermore, the new series of precatalysts are air-, moisture and thermally-stable and display good solubility in common organic solvents. The use of precatalysts ensures the efficient generation of the active catalytic species and allows one to accurately adjust the ligand:palladium ratio. The ligands, precatalysts and methodology developed in the Buchwald group are user friendly and have rendered previously difficult cross couplings reactions, much easier to achieve.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service