Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

687227

Sigma-Aldrich

1H-Indole-2-methanol

90%

Synonym(s):

2-(Hydroxymethyl)indole, NSC 165230

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

72-78 °C

SMILES string

OCc1cc2ccccc2[nH]1

InChI

1S/C9H9NO/c11-6-8-5-7-3-1-2-4-9(7)10-8/h1-5,10-11H,6H2

InChI key

XEEANGGQJOWRTG-UHFFFAOYSA-N

Application

Reactant for the synthesis of:
  • Oxazino[4,3-a]indoles
  • 2-arylsulfonylmethyl-3-piperazinylmethylindole derivatives as 5-HT6 receptor ligands
  • SR13668 designed to mimic the unique anticancer mechanisms of dietary indole-3-carbinol to block Akt signaling
  • Azido- and isothiocyanato-substituted indoles as melatoninergic agents
Useful starter in indole chemistry.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 13, 1810-1810 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service