Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

679267

Sigma-Aldrich

Allyltrimethoxysilane

≥98%, deposition grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2Si(OCH3)3
CAS Number:
Molecular Weight:
162.26
Beilstein:
2350745
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

146-148 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CC=C)(OC)OC

InChI

1S/C6H14O3Si/c1-5-6-10(7-2,8-3)9-4/h5H,1,6H2,2-4H3

InChI key

LFRDHGNFBLIJIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cary A Kuliasha et al.
Langmuir : the ACS journal of surfaces and colloids, 36(1), 379-387 (2019-12-13)
Effectively negating the deleterious impact of marine biofouling on the world's maritime fleet in an environmentally conscientious manner presents a difficult challenge due to a variety of factors including the complexity and diversity of fouling species and the differing surface

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service