Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667994

Sigma-Aldrich

3-Methyl-2-furoic acid

97%

Synonym(s):

3-Methylfuran-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

134-138 °C

SMILES string

Cc1ccoc1C(O)=O

InChI

1S/C6H6O3/c1-4-2-3-9-5(4)6(7)8/h2-3H,1H3,(H,7,8)

InChI key

BNYIQEFWGSXIKQ-UHFFFAOYSA-N

Application

Used in a palladium-catalyzed cross-coupling between heteroaryl carboxylic acids and aryl bromides leading to arylated heterocycles. Also used in a palladium-catalyzed asymmetric hydrogenation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Catal., 226, 52-52 (2003)
Pat Forgione et al.
Journal of the American Chemical Society, 128(35), 11350-11351 (2006-08-31)
Aryl-substituted five-membered heteroaromatics have attracted great interest over the past years due to their presence in a large number of pharmaceuticals and natural products. Recently, an advance in the preparation of these scaffolds was achieved by employing a C-H functionalization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service