Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

663115

Sigma-Aldrich

2-Iodoadenosine

97%

Synonym(s):

Butyltrimethylammonium-bis(trifluoromethanesulfonyl)imide, N,N,N-Trimethyl-1-butanaminium bis[(trifluoromethyl)sulfonyl]imide, N,N,N-Trimethyl-N-propylammonium bis(trifluoromethylsulfonyl)amide, N-Trimethyl-N-butylammonium bis(trifluoromethanesulfonyl)imide, Trimethylbutylammonium bis(trifluoromethanesulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12IN5O4
CAS Number:
Molecular Weight:
393.14
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

200 °C (dec.)

SMILES string

Nc1nc(I)nc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H12IN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1

InChI key

MGEBVSZZNFOIRB-UUOKFMHZSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed cross-coupling of 2-iodoadenosine with terminal alkynes: synthesis and biological activities of 2-alkynyladenosines.
A Matsuda et al.
Chemical & pharmaceutical bulletin, 33(4), 1766-1769 (1985-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service