Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

617253

Sigma-Aldrich

Dimethyl-d6 phthalate

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D6H4O4
CAS Number:
Molecular Weight:
200.22
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

isotopic purity

98 atom % D

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.214 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])OC(=O)c1ccccc1C(=O)OC([2H])([2H])[2H]

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3/i1D3,2D3

InChI key

NIQCNGHVCWTJSM-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service