Skip to Content
MilliporeSigma
All Photos(1)

Documents

606006

Sigma-Aldrich

Sodium L-lactate-2,3-13C solution

45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Synonym(s):

L-Lactic acid-2,3-13C2 sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CH(OH)CO2Na
CAS Number:
Molecular Weight:
114.05
UNSPSC Code:
12352001
PubChem Substance ID:

description

Packaged as gram of solution, 45-55%(w/w)

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

concentration

45-55 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

mp

163-165 °C (lit.)

mass shift

M+2

storage temp.

−20°C

SMILES string

[Na+].[13CH3][13CH](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/i1+1,2+1;

InChI key

NGSFWBMYFKHRBD-AWQJXPNKSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service