Skip to Content
MilliporeSigma
All Photos(1)

Documents

590711

Sigma-Aldrich

4-Chloro-2-methylaniline

98%

Synonym(s):

2-Amino-5-chlorotoluene, 4-Chloro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
878505
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.583 (lit.)

bp

241 °C (lit.)

mp

24-27 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(Cl)ccc1N

InChI

1S/C7H8ClN/c1-5-4-6(8)2-3-7(5)9/h2-4H,9H2,1H3

InChI key

CXNVOWPRHWWCQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chloro-2-methylaniline may be used in the synthesis of the following:
  • 2,8-dichloro-4,10-dimethyl-6H,12H-5,11-methanodibenzo[b,f]-diazocine via reaction with paraformaldehyde in toluene
  • N-allylated derivative via palladium-catalyzed selective monoallylation with allyl alcohol
  • N-(4-chloro-2-methylphenyl)benzamide via reaction with benzoyl chloride

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Direct palladium-catalyzed selective monoallylation of
anilines using allylic alcohols?
Yang C-S, et al.
Tetrahedron Letters, 41(36), 7097-7100 (2000)
"Synthesis of halogen substituted analogues of Troger?s base"
Jensen J and Warnmark K
Synthesis, 2001(12), 1873-1877 (2001)
"Derivatives from Isoselenocyanates: Synthesis of 2-Phenyl-6H-[5, 1, 3] benzoselenadiazocine"
Atanassov.KP, et al.
Helvetica Chimica Acta, 87(06), 1452-1466 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service