Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590096

Sigma-Aldrich

Dimethyl sulfate-13C2,d6

99 atom % 13C, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(13CD3O)2SO2
CAS Number:
Molecular Weight:
134.15
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

99% (CP)

technique(s)

protein expression: suitable

bp

188 °C (lit.)

mp

-32 °C (lit.)

density

1.417 g/mL at 25 °C

mass shift

M+8

SMILES string

[2H][13C]([2H])([2H])OS(=O)(=O)O[13C]([2H])([2H])[2H]

InChI

1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3/i1+1D3,2+1D3

InChI key

VAYGXNSJCAHWJZ-KLFIIISESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service