Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57903

Sigma-Aldrich

(Iodomethyl)trimethylsilane

≥99.0% (GC)

Synonym(s):

(Trimethylsilyl)methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2I
CAS Number:
Molecular Weight:
214.12
Beilstein:
1731485
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.491 (lit.)
n20/D 1.491

bp

139-141 °C (lit.)

density

1.443 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)CI

InChI

1S/C4H11ISi/c1-6(2,3)4-5/h4H2,1-3H3

InChI key

VZNYXGQMDSRJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for the preparation of 2-(trimethylsilyl)ethyl-triphenylphosphonium iodide used for the synthesis of allyl silanes; Used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Vedejs et al.
The Journal of Organic Chemistry, 48, 4773-4773 (1983)
E. Vedejs et al.
The Journal of Organic Chemistry, 50, 2170-2170 (1985)
D. Seyferth et al.
The Journal of Organic Chemistry, 42, 3104-3104 (1977)
I. Fleming et al.
Synthesis, 446-446 (1979)
Aleksandra Sadowska et al.
Frontiers in pharmacology, 11, 952-952 (2020-07-28)
Painful intervertebral disc (IVD) degeneration is an age-related process characterized by reduced tissue osmolarity, increased catabolism of the extracellular matrix, and elevated levels of pro-inflammatory molecules. With the aging population and constantly rising treatment costs, it is of utmost importance

Articles

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Review SET mechanism in photoredox catalysis for sustainable reactions and rearrangement of oxaziridines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service