Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

574023

Sigma-Aldrich

Cesium perchlorate

99.9% trace metals basis

Synonym(s):

Perchloric acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsClO4
CAS Number:
Molecular Weight:
232.36
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: oxidant

mp

250 °C (dec.) (lit.)

SMILES string

[Cs+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Cs/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

WKDKOOITVYKILI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Investigations of ionic conductivity in orientationally disordered phases

Used for:
  • Long-duration optical transmission of information with infofuses
  • Modifier of polymer electrolyte/luminescent conjugated polymer mixed ionic-electronic conductors
  • Studies involving pH-dependent mass and volume changes of polypyrrole / poly(styrene sulfonate)
  • Cationic effects in polymer light-emitting electrochemical cells

Reactant for investigating metal salt effects on morphology of silica nanomaterials prepared by polymerization on hydrogels

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service